Learn More
Aqua Solutions SULFURIC ACID 0.02 NORMAL 4LT
SDP
Supplier:  Aqua Solutions 90204L
Encompass_Preferred
Sulfuric Acid; 0.02N; Clear, liquid; 4L; M.P. ~2 deg.C; B.P. 100 deg.C; Percent Volatile by Volume:> 99%; CAS: 7664-93-9; Use with adequate ventilation
Catalog No. NC9123722
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov