Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 0.2 N Solution, Spectrum™ Chemical
SDP

Catalog No. 18607142 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-142 4 L Poly Bottle
18-607-144 500 mL Poly Bottle
18-607-141 1 L Poly Bottle
18-607-143 20 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-142 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S6204LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S-620, 7664-93-9

Spectrum™ Chemical Sulfuric Acid, 0.2 N Solution - Spectrum utilizes the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Concentration 0.2 N ±0.005
CAS Max % 1 to <3%, 80 to 100%
Packaging Poly Bottle
Quantity 4 L
Grade Ungraded
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.