Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, 1.0N Standardized Solution

Catalog No. aa35655k2
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA35655K2 1 L
AA35655K7 4 L
AA35655M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA35655K2 Supplier Thermo Scientific Chemicals Supplier No. 035655K2
Only null left
Add to Cart

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid, 1.0N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Physical Form Liquid
Solubility Miscible with water
Quantity 1 L
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 5%; Water: 95%
Chemical Name or Material Sulfuric acid
Hazard Category H290-H315-H318-H335
Precautionary Statement P234-P261-P264b-P271-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P362-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.