Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 1.8 N, with Catalyst, Ricca Chemical
SDP

Catalog No. R830800020E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R830800020E 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R830800020E Supplier Ricca Chemical Company Supplier No. R830800020E1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory grade 1.8 N sulfuric acid solution with catalyst.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9 , 10034-96-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Appearance of Solution Colorless Liquid
Concentration 1.79 to 1.81 N
CAS Max % 91.51, 8.49, 0.0037
Packaging Hard Pack
Quantity 20 L
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Sulfuric Acid
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.