Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 18.0 N, Ricca Chemical
SDP

Catalog No. R83431804A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R83431804A 4 L Natural Poly Bottle
834318-16 500 mL Natural Poly Bottle
834318-32 1 L Natural Poly Bottle
834318-5PT 2.5 L Safety-coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R83431804A Supplier Ricca Chemical Company Supplier No. R83431804A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Appearance of Solution Colorless Liquid
Color Colorless
CAS Min % 51.0, 47.0
CAS Max % 53.0, 48.92
Concentration 17.95 to 18.05 N
Packaging Natural Poly Bottle
Quantity 4 L
Physical Form Liquid
Synonym oil of vitriol
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Sulfuric Acid
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.