Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 18 M, ACS Grade, Innovating Science™
SCI_ED

Catalog No. S02444 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02444 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING! : This product can expose you a chemical, Strong inorganic acid mists containing sulfuric acid, which is known to the State of California to cause cancer.
Catalog No. S02444 Supplier Aldon Corporation Supplier No. IS28250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING! : This product can expose you a chemical, Strong inorganic acid mists containing sulfuric acid, which is known to the State of California to cause cancer.

High-purity ACS grade sulfuric acid for laboratory and analytical applications.

Chemical Identifiers

CAS 7732-18-5
Molecular Weight (g/mol) 98.08

Specifications

Concentration 18 M
Packaging Amber Glass
Quantity 500 mL
Percent Purity 95.0% to 98.0%
Grade Laboratory
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.