Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 20% (v/v), Reagent Grade, Ricca Chemical
SDP

Catalog No. R816500020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R816500020F 20 L Cubitainer
8165-16 500 mL Natural Poly Bottle
8165-32 1 L Natural Poly Bottle
8165-1 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R816500020F Supplier Ricca Chemical Company Supplier No. R816500020F
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Appearance of Solution Colorless Liquid
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 20 L
Synonym oil of vitriol
Percent Purity 19.97 to 20.03%
Grade Reagent
Chemical Name or Material Sulfuric Acid, 20% (v/v)
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.