Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 20% w/v, Reagents

Catalog No. 01333879
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Cubitainer
HDPE Winpak
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-333-879 1 L Poly Bottle
01-333-878 20 L HDPE Winpak
01-333-877 20 L Cubitainer
01-333-876 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-333-879 Supplier Reagents Holdings Llc Supplier No. CS1017001A
Only null left
Add to Cart
Add to Cart

Sulfuric Acid, 20% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.1251
Quantity 1 L
Physical Form Liquid
Percent Purity 20%
Grade Reagent
Chemical Name or Material Sulfuric Acid, 20% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.