Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 25%, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1007169025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 L
Packaging:
Plastic Container
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1007169025 25 L Plastic Container
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1007169025 Supplier MilliporeSigma Supplier No. 1007169025
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric acid for analytical applications, providing high-purity chemical reagent performance.

Recommended Storage

Storage temperature: no restrictions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Boiling Point 103°C (1013hPa)
Density 1.18 g/3 (20 °C)
Packaging Plastic Container
Quantity 25 L
Physical Form Liquid
Solubility Information Soluble at 68°F (20°C)
Formula Weight 98.08
Percent Purity ≥25.0%
Grade Analytical
Chemical Name or Material Sulphuric Acid
Show More Show Less
EINECSNumber 231-639-5
RTECSNumber WS5600000
Recommended Storage Storage temperature: no restrictions.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.