Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 3.00 N, Ricca Chemical
SDP

Catalog No. R8320000500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R8320000500 500 mL Natural Poly Bottle
8320-32 1 L Natural Poly Bottle
8320-1 4 L Natural Poly Bottle
8320-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R8320000500 Supplier Ricca Chemical Company Supplier No. 832016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7732-18-5 | H2O4S | 98.07 g/mol

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Odor Odorless
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Assay Percent Range 3.000 ± 0.001 N at 20°C
Quantity 500 mL
Physical Form Liquid
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Chemical Name or Material Sulfuric Acid, 3.00 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.