Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 50% w/w, Reagents

Catalog No. 01333870
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-870 500 mL
01-333-871 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-870 Supplier Reagents Holdings Llc Supplier No. CS102200500A
Only null left
Add to Cart
Add to Cart

Sulfuric Acid, 50% w/w, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 120°C
Packaging Poly Bottle
Specific Gravity 1.3977
Quantity 500 mL
Physical Form Liquid
Percent Purity 50%
Grade Reagent
Chemical Name or Material Sulfuric Acid, 50% w/w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.