Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 66 °Be, Technical, Approx. 93%, Spectrum™ Chemical
SDP

Catalog No. 18607156 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-156 4 x 2.5 L Glass Bottle
18-607-153 500 mL Amber Glass Bottle
18-607-155 2.5 L Glass Bottle
18-607-157 4 L Amber Glass Bottle
18-607-158 4 x 4 L Amber Glass Bottle
18-607-152 20 L Poly Drum
18-607-154 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-156 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S17352.5LTGLCS4
Only null left
Add to Cart
Add to Cart

S1735, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, 66 °Be, Technical is an extremely corrosive acid that comes as a yellowish slightly viscous liquid. It is soluble in water and is a diprotic acid. It has very strong corrosive, dehydrating and oxidizing properties as well as being hygroscopic. It uses range from detergent production, antifreeze, pharmaceuticals and fertilizers to cellophane, lubricants and production of explosives. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 100%
Packaging Glass Bottle
Quantity 4 x 2.5 L
Percent Purity ∼93%
Grade Technical
Chemical Name or Material Sulfuric Acid
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.