Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 66% v/v, Reagents

Catalog No. 01333529
Click to view available options
:
Quantity: 500 mL

Catalog No. 01-333-529

Supplier: Reagents Holdings Llc CS203500500A

Only null left
Add to Cart
Add to Cart

Sulfuric Acid, 66% v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9, 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 185°C
Packaging Poly Bottle
Specific Gravity 1.5572
Quantity 500 mL
Physical Form Liquid
Percent Purity 66%
Grade Reagent
Chemical Name or Material Sulfuric Acid, 66% v/v
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.