Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, 75% v/v aq. soln.

Catalog No. AA42555K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42555K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA42555K2 Supplier Thermo Scientific Chemicals Supplier No. 042555K2
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid has an useful applications in domestic acidic drain cleaner, as a drying agent, electrolyte in lead-acid batteries and various cleaning agents. It is also useful for mineral processing, fertilizer manufacturing, oil refining, waste water processing, and chemical synthesis. The sulfuric acid doped poly (m-aminophenol) is used as aliphatic alcohol vapor sensor material. The catalytic decomposition of sulfuric acid on mixed Cr/Fe oxide, shows an useful application in sulfur-iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Appearance Colorless to pale yellow liquid
Odor Acrid
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Assay Percent Range 75% v/v aqueous
Quantity 1 L
Physical Form Liquid
Solubility Information Miscible with water,acetic acid and alcohol.
Formula Weight 98.08
Concentration or Composition (by Analyte or Components) Sulfuric acid: 84.7%; Water: 15.3%
Chemical Name or Material Sulfuric acid
Hazard Category H314-H335-H350
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.