Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, Environmental Grade, 93-98%

Catalog No. aa38751ap
Change view
Click to view available options
Quantity:
500 mL
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA38751AP 500 mL
AA38751K4 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA38751AP Supplier Thermo Scientific Chemicals Supplier No. 038751AP
Only null left
Add to Cart

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Sulfuric acid has a strong dehydrating agent and is applied in many applications like the domestic acidic drain cleaner, electrolyte in lead-acid batteries and in various other cleansing agents. It is also useful for mineral processing, fertilizer manufacturing, petroleum purification, wastewater processing, and chemical synthesis. The catalytic decomposition of sulfuric acid when mixed with Cr/Fe oxides, shows a useful application in the sulfur iodine cycle for hydrogen production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Appearance Colorless to pale yellow liquid
Color Colorless to Yellow
Density 1.84
Odor Acrid
Assay Percent Range 93 - 98%
Quantity 500 mL
Physical Form Liquid,single sub-boiling quartz distillation
Solubility Information Miscible in water.
Formula Weight 98.08
Grade Environmental
Chemical Name or Material Sulfuric acid, Environmental Grade
Show More Show Less
Hazard Category H314-H330-H350
Precautionary Statement P201-P202-P260-P264b-P271-P280-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P363-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-639-5
RTECSNumber WS5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.