Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 96.0%, Ultrapur, MilliporeSigma™

Catalog No. M1015160250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1015160250 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1015160250 Supplier MilliporeSigma Supplier No. 1.01516.0250
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity grade sulfuric acid suitable for trace element analysis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.08

Specifications

pH 0.3 (49 g/L, H2O, 25°C)
Melting Point −20°C
Density 1.84 g/cm3 (at 20°C)
Packaging Plastic Bottle
Vapor Pressure 0.0001 hPa (20°C)
Assay Percent Range ≥ 96%
Quantity 250 mL
Percent Purity 96%
Grade Not Available
Chemical Name or Material Sulfuric Acid
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.