Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 98.0%, EMPROVE™ ESSENTIAL Ph Eur, BP, JPE, NF, MilliporeSigma™

Catalog No. M1007131000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1007131000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1007131000 Supplier MilliporeSigma Supplier No. 1.00713.1000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3 (49 g/L, H2O, 25°C)
Melting Point −20°C
Boiling Point 335°C (1013 hPa)
Density 1.84 g/cm3 (at 20°C)
Quantity 1 L
Packaging Glass Bottle
Assay Percent Range 95.0 to 98.0%
Vapor Pressure 0.0001 hPa (20°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.