Learn More
Specifications
Specifications
| Color | Colorless |
| CAS Min % | 95.0 |
| CAS Max % | 98.0 |
| Heavy Metals (as Pb) | max. 0.001% |
| Packaging | Amber Glass Bottle |
| Reducing Substances | Pass |
| Quantity | 1 L |
| Physical Form | Liquid |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| Percent Purity | ≥85% |
| Show More |
Safety and Handling
| ShelfLife | 36 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
