Learn More
Description
- For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; NOTE: Intended for nonfuel end purposes only
Specifications
Specifications
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.84kg |
| Quantity | 2.5 L |
| Formula Weight | 98.08 |
| Grade | Meets ACS reagent chemical requirements |
| Chemical Name or Material | Sulfuric Acid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.