Learn More
Specifications
Specifications
| Color | Colorless |
| Concentration | 1.000N ± 0.005N |
| CAS Max % | 95.24, 4.76 |
| Density | 1.03g/mL |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1.03g/mL |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Sulfur compounds; Thermal decomposition generates: Corrosive vapors |
| Quantity | 1 L |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves/eye protection. Wash exposed skin thoroughly after handling. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/docto |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.