Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Certified, 1.000N ±0.005N (0.5M), LabChem™

Catalog No. LC257704
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC257704 4 L
LC257701 500 mL
LC257702 1 L
LC257706 10 L
LC257705 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC257704 Supplier LabChem, Inc. Supplier No. LC257704
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 1.000N ± 0.005N
CAS Max % 95.24, 4.76
Density 1.03g/mL
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.03g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Quantity 4 L
Physical Form Liquid
Linear Formula H2SO4
UN Number UN2796
Solubility Information Soluble in water
Formula Weight 98.08
Grade Certified
Chemical Name or Material Sulfuric Acid, 1.0N (0.5M)
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.