Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid concentrate, For 1L standard solution, 0.05 M H2SO4 (0.1N), Solstice

Catalog No. 60046055 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-055 6 x 1 Ea.
60-016-26 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-055 Supplier Solstice Supplier No. 320436X1EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.05 M H2SO4 (0.1N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % ≥90.0000%, ≥5.0000%
CAS Max % ≤100.0000%, <10.0000%
Density 1.05 g/cm3
Packaging Ampoule
Quantity 6 x 1 Ea.
Physical Form Concentrate
Linear Formula H2SO4
UN Number UN2796
Beilstein 2037554
Formula Weight 98.08
Grade Not Available
Chemical Name or Material Sulfuric acid concentrate, For 1L standard solution, 0.05 M H2SO4 (0.1N)
Show More Show Less
Hazard Statement P305 + P351 + P338
Precautionary Statement H290-H315-H319
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sulfuric acid
EINECSNumber 231-639-5
Recommended Storage Room Temp
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.