Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 49.04g H2SO4, J.T. Baker™

Catalog No. 02004369 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 Pk.
Packaging:
Poly Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-369 1 Pk. Poly Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-369 Supplier Avantor Supplier No. 470001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent concentrate designed for convenient dilution to 1 N aqueous solution.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Packaging Poly Ampoule
Specific Gravity 1L = 1.40kg
Quantity 1 Pk.
Grade Laboratory
Chemical Name or Material Sulfuric Acid, DILUT-IT™ Analytical Concentrate, 1N, 1 equiv. = 49.04g H2SO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.