Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Electronic/Cleanroom Grade, 95-97%, Spectrum™ Chemical
SDP

Catalog No. 18607159 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-607-159 1 gal.
18-607-160 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-607-159 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S17421GL
Only null left
Add to Cart
Add to Cart

S1742, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, Electronic/Cleanroom Grade is an extremely corrosive acid that comes as a yellowish slightly viscous liquid. It is soluble in water and is a diprotic acid. It has very strong corrosive, dehydrating and oxidizing properties as well as being hygroscopic. Spectrum’s ELECTRONIC/CLEANROOM GRADE products are suitable for use in electronic component manufacturing. These materials meet or exceed the specifications as listed in Reagent Chemicals published by the American Chemical Society.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 1 gal.
Percent Purity 95 to 97%
Grade Electronic/Cleanroom
Chemical Name or Material Sodium Sulfathiazole, Sesquihydrate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.