Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, FCC, 95-98%, Spectrum™ Chemical
SDP

Catalog No. 18607167 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
200 L
Packaging:
Amber Glass Bottle
Drum
Glass Bottle
Poly-Coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-167 200 L Drum
18-607-165 500 mL Amber Glass Bottle
18-607-162 2.5 L Glass Bottle
18-607-161 2.5 L Poly-Coated Glass Bottle
18-607-166 6 x 500 mL Amber Glass Bottle
18-607-164 4 x 2.5 L Glass Bottle
18-607-163 4 x 2.5 L Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-167 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1736200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1736, 7664-93-9, H2SO4

Spectrum™ Chemical Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 95 to 98%
Grade FCC
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.