Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, Solstice

Catalog No. 60090563 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-563 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-563 Supplier Solstice Supplier No. 40254H2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Semiconductor grade ( 17831), 95-97%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Boiling Point 310°C (1.013 hPa)
CAS Min % ≥1.00%, ≥50.00%
CAS Max % <5.00%, ≤100.00%
Packaging Plastic Bottle
Quantity 2.5 L
Physical Form Liquid
Beilstein 2037554
Percent Purity ≥95%
Grade Semiconductor grade acid
Chemical Name or Material Sulfuric Acid
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.