Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric acid, Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%, Solstice

Catalog No. 6001593 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Glass Bottle
Plastic Bottle
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-015-93 1 L Glass Bottle
60-015-95 2.5 L Glass Bottle
60-015-96 5 L Plastic Bottle
60-045-850 6 x 1 L Plastic Bottle
60-045-848 4 x 2.5 L Glass Bottle
60-045-847 4 x 2.5 L Plastic Bottle
60-045-849 4 x 5 L Plastic Bottle
60-015-92 1 L Plastic Bottle
60-015-94 2.5 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 60-015-93 Supplier Solstice Supplier No. 307431LGL
Add to Cart
Add to Cart

Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Density 1.840g/mL at 25°C
Packaging Glass Bottle
Residue after Ignition ≤5ppm
Vapor Pressure 1mmHg (146°C)
Quantity 1 L
Linear Formula H2SO4
UN Number 1830
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity 95 to 97%
Grade ACS/puriss. p.a.
Chemical Name or Material Sulfuric Acid, Reag. Ph. Eur. reag. ISO
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H290-H314
EINECSNumber 231-639-5
RTECSNumber WS5600000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.