Learn More
Sulfuric acid solution, Volumetric, Reag. Ph. Eur., 0.5 M H2SO4 (1.0N), Solstice
Shop All Solstice Research Chemicals ProductsDescription
Solstice's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
| Concentration | 0.5M |
| Packaging | VOLPAC Container |
| Quantity | 10 L |
| Linear Formula | H2SO4 |
| UN Number | UN3264 |
| Beilstein | 2037554 |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| Formula Weight | 98.08g/mol |
| Grade | Reagent Ph. Eur. |
| Chemical Name or Material | Sulfuric acid solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.