Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid Solution, 92.8%, for Babcock Milk Test, MilliporeSigma™

Catalog No. 50948029 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-948-029 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-948-029 Supplier MilliporeSigma Supplier No. SX1244P4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Optimized sulfuric acid solution specifically formulated for use in the Babcock milk test.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Weight (g/mol) 98.08

Specifications

pH 0.3 (49 g/L, H2O, 25°C)
Melting Point −20°C
Boiling Point 335°C (1013 hPa)
Packaging Plastic Bottle
Vapor Pressure 0.0001 hPa (20°C)
Assay Percent Range 90.5 to 92.8%
Quantity 4 L
Percent Purity 90.5 to 92.8%
Chemical Name or Material Sulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.