Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, 10% (w/w), Ricca Chemical
SDP

Catalog No. 815325 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
55 gal.
Packaging:
Cubitainer
Natural Poly Bottle
Poly Drum
Safety-coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
815325 10 L Cubitainer
815332 1 L Natural Poly Bottle
81535PT 2.5 L Safety-coated Glass Bottle
81535 20 L Cubitainer
815355 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 815325 Supplier Ricca Chemical Company Supplier No. 81532.5
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 10% (w/w)
CAS Min % 89.0, 9.0
CAS Max % 91.0, 11.0
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Quantity 10 L
Physical Form Liquid
Percent Purity 10% W/W
Grade Laboratory
Chemical Name or Material Sulfuric Acid Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.