Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, TraceSELECT™ Ultra, ≥95% (T), Solstice

Catalog No. 60047545 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Packaging:
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-545 250 mL Poly bottle
60-047-546 500 mL Poly bottle
60-047-544 1 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-545 Supplier Solstice Supplier No. 77239250ML
Add to Cart
Add to Cart

TraceSELECT™ Ultra, for ultratrace analysis,≥95%

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH <1
Melting Point -18°C
Boiling Point 203°C
Density 1.840g/mL at 25°C, 1.83g/mL at 20°C
Odor Odorless
Packaging Poly bottle
Vapor Pressure 1mmHg (146°C)
Quantity 250 mL
Physical Form Liquid
Linear Formula H2SO4
UN Number UN1830
Beilstein 2037554
Formula Weight 98.08g/mol
Percent Purity ≥95% (T)
Grade TraceSELECT™ Ultra
Chemical Name or Material Sulfuric acid
Show More Show Less
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
DOTInformation 8 / PGII
RTECSNumber 231-639-5
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.