Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuric Acid, Ultratrace, 93-98%, Spectrum™ Chemical
SDP

Catalog No. 18614299 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
PTFE Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-299 500 mL PTFE Bottle
18-614-298 1 L PTFE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-299 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1072500MLTF
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Max % 1
Packaging PTFE Bottle
Quantity 500 mL
Percent Purity 93 to 98%
Grade High Purity Solvent
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.