Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfurous Acid, 6% ACS Reagent, Reagents

Catalog No. 01338158
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-158 500 mL
01-338-160 1 L
01-338-159 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-338-158 Supplier Reagents Holdings Llc Supplier No. C1115500500C
Add to Cart
Add to Cart

Sulfurous Acid, 6% ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7782-99-2 , 7732-18-5
Molecular Formula H2O3S
Molecular Weight (g/mol) 82.07
InChI Key LSNNMFCWUKXFEE-UHFFFAOYSA-N
Synonym Sulfur dioxide solution, Sulphurous Acid
IUPAC Name sulfurous acid
SMILES OS(O)=O

Specifications

pH <7
Color Colorless
Boiling Point 35°C
Packaging Glass Bottle
Specific Gravity 1.03
Quantity 500 mL
Physical Form Liquid
Grade ACS
Chemical Name or Material Sulfurous Acid, 6% ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.