Learn More
Medchemexpress LLC Suloctidil | 54767-75-8 | 100.0% | 337.56 | C20H35NOS | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB123710MG
Encompass_Preferred
Suloctidil is a sulfur-containing aminoalcohol used as a Na+/K+ ATPase inhibitor in pharmacological research and in vitro studies. It is supplied for small-scale laboratory work and is characterized by high analytical purity and documented solubility in common laboratory solvents.
Catalog No. 50-002-97434