Learn More
Description
- This is an in vitro semi-quantitative method for antimicrobial susceptibility of clinical isolates tested on agar media using overnight incubation
- The 250-test box contains 5 cartridges of 50 discs, each cartridge individually packed in a desiccant envelope
- These are discs with special features, that are impregnated with antibiotic and used for the susceptibility test according to the Kirby-Bauer antibiotic testing (KB testing or disc diffusion antibiotic sensitivity testing)
Specifications
Specifications
| Product Type | Antimicrobial Susceptibility Disc, Antibiotic Test Disc |
| Target Organism Class | Gram-negative Microorganisms |
| Test Type | Antimicrobial Susceptibility, Antibiotic Susceptibility Sensitivity, Microbial Susceptibility |
| Species | Enterobacterales |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
