Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Sulprostone, EP1/ EP3 Receptor Agonist, >98% Purity, 1 mg

Catalog No. 502707140
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7140 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7140 Supplier Abcam Supplier No. AB1209071MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Selective PGE2 analog and EP1/ EP3 receptor agonist with >98% purity.

  • Molecular weight: 465.65 g/mol
  • Purity: >98%
  • Potency: Selective EP1/ EP3 receptor agonist (Ki values are 21 and 0.6 nM, respectively)
  • Chemical class: PGE2 analog
  • Storage buffer: Constituents: 99.5% Methyl acetate, 0.5% Sulprostone
  • Form: Liquid
  • Storage temperature: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule EP1 Receptor, EP3 Receptor
Form Liquid
Buffer 99.5% methyl acetate, 0.5% sulprostone
Content And Storage Store at -20°C.
Formulation 99.5% Methyl acetate, 0.5% Sulprostone
Quantity 1 mg
For Use With (Application) EP1-EP3 receptor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.