Learn More
Invitrogen™ SULT1A4 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SULT1A3.
Specifications
Specifications
| Antigen | SULT1A4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | SULT1A4 |
| Gene Accession No. | P0DMN0 |
| Gene Alias | aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; HAST3; monoamine-sulfating phenol sulfotransferase; M-PST; Placental estrogen sulfotransferase; SLX1B-SULT1A4; ST1A3; ST1A3/ST1A4; ST1A4; STM; sulfokinase; Sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 4; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4; sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; Thermolabile phenol sulfotransferase; TL-PST |
| Gene Symbols | SULT1A4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.