Learn More
Invitrogen™ SULT1B1 Monoclonal Antibody (724429)

Description
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Specifications
Specifications
| Antigen | SULT1B1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 724429 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | SULT1B1 |
| Gene Accession No. | O43704 |
| Gene Alias | DOPA/tyrosine sulfotransferase; ST1B1; ST1B2; St2b2; sulfotransferase 1B1; sulfotransferase 1B2; sulfotransferase family 1B member 1; sulfotransferase family 1B, member 1; sulfotransferase family cytosolic 1B member 1; sulfotransferase family, cytosolic, 1B, member 1; SULT1B1; SULT1B2; Thyroid hormone sulfotransferase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.