Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Sumatriptan Succinate, 5-HT1 Receptor Agonist, Solid, >99% Purity, 50 mg
Description
- Molecular weight: 413.49 g/mol
- Purity: >99%
- Mechanism: 5-HT1 receptor agonist (Ki values: 17 nM at 5-HT1D, 27 nM at 5-HT1B, and 100 nM at 5-HT1A receptors)
- Affinity: 5-HT1F receptor (EC50 = 247 nM)
- Biological activity: Reduces vascular inflammation associated with migraine
- Form: Solid
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 5-HT1 Receptor |
| Buffer | 100% Sumatriptan succinate |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Shipping Condition | Ambient |
| Formulation | 100% Sumatriptan succinate |
| Quantity | 50 mg |
| For Use With (Application) | 5-HT1 receptor agonist, Migraine research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?