Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SUMO2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. p-7184284
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111147 100 μL
PIPA5111266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111147 Supplier Invitrogen™ Supplier No. PA5111147
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TDKPAQLEME DEDTIDVFQQ PTGGV Highest antigen sequence indentity to the following orthologs: Mouse - 92%, Rat - 92%.

SUMO2 is a member of the SUMO (small ubiquitin-like modifier) protein family. This protein family functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. However, unlike ubiquitin which targets proteins for degradation, this protein is involved in a variety of cellular processes, such as nuclear transport, transcriptional regulation, apoptosis, and protein stability. In vertebrates, three members of the SUMO family have been described, SUMO 1 and the functionally distinct homologues SUMO 2 and SUMO 3. SUMO modification sites present in the N terminal regions of SUMO 2 and SUMO 3 are utilized by SAE1/SAE2 (SUMO E1) and Ubc9 (SUMO E2) to form polymeric chains of SUMO 2 and SUMO 3 on protein substrates, a property not shared by SUMO 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUMO2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SUMO2
Gene Accession No. P61956
Gene Alias HSMT3; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 2 L homeolog; small ubiquitin-like modifier 3; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 2-A; small ubiquitin-related modifier 3; SMT3 homolog 1; SMT3 homolog 2; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 supressor of mif two 3 homolog 2; Smt3A; Smt3b; SMT3H1; SMT3H2; Sumo2; SUMO-2; sumo2.L; SUMO-2-A; sumo2-a; sumo2-b; SUMO3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; XELAEV_18044053mg
Gene Symbols SUMO2
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human SUMO2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6613
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.