Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SUMOylation Assay Kit, Enzyme Activity, 20 Test

Catalog No. 502583435
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3435 20 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3435 Supplier Abcam Supplier No. AB13947020TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to assay SUMOylation in cell and tissue extracts, cited in over 15 publications.

  • Assay type: Enzyme activity
  • Sample types: Purified protein
  • Usage: Cited in > 15 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Product Type Western Blot Kit
Format Kit
No. of Reactions 20 Tests
Target Molecule SUMOylation
Sample Type Purified protein
Quantity 20 Tests
For Use With (Application) SUMOylation assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.