Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ SUN1, Mouse, Unlabeled, Clone: 12.10F,

Catalog No. MABT892MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABT892MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT892MI Supplier MilliporeSigma™ Supplier No. MABT892
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SUN domain-containing protein 1 (UniProt Q9D666; also known as Protein unc-84 homolog A, Sad1/unc-84 protein-like 1) is encoded by the Sun1 (also known as Unc84a) gene (Gene ID 77053) in murine species. SUN1 is a component of the SUN-protein-containing complexes known as LINC (linker of nucleoskeleton and cytoskeleton), which link the nucleoskeleton and cytoskeleton structures together in a scaffold like fashion. SUN1 is required for interkinetic nuclear migration and nucleokinesis as well as nucleus-centrosome coupling during radial neuronal migration and glial migration in the cortex. SUN1 also is important during meiosis and is required for proper gametogenesis. It also participates in anchoring chromosome movement in the prophase of meiosis and in selective gene expression of coding and non-coding RNAs required for gametogenesis. Additionally, it has a vital role in the distribution of nuclear pore complexes on the nuclear membrane. Interestingly, LINC complexes are also critical for vital functions such as hearing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SUN1
Applications Western Blot
Classification Monoclonal
Clone 12.10F
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mous IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Sun1, Unc84a
Gene Accession No. Q9D666
Host Species Mouse
Immunogen GST-tagged recombinant fusion protein composed of murine SUN1 regions encoded by exons 6, 8, and 10.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_077771
Test Specificity Clone 12.10F was raised against a fusion construct composed of murine SUN1 exon 6-, 8-, and 10-encoded regions that are present in murine spliced isoforms 1, 3, and 4 reported by UniProt (Q9D666). The epitope location among these three regions has not been mapped and reactivity toward murine spliced isoform 3 is expected only if the epitope resides within exon 1- or 4-encoded region.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.