Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SUN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501735699 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5699 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5699 Supplier Proteintech Group Inc Supplier No. 275561AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UNC84B may form a physical interaction between the nuclear envelope and the centrosome.

Specifications

Antigen SUN2
Applications Western Blot
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SUN2
Gene Accession No. Q8BJS4, Q9UH99
Gene Alias FRIGG, KIAA0668, Protein unc 84 homolog B, Rab5 interacting protein, Rab5IP, Sad1/unc 84 protein like 2, SUN2, unc 84 homolog B (C. elegans), UNC84B
Gene Symbols Sun2
Host Species Rabbit
Immunogen SUN2 Fusion Protein Ag16077
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223697, 25777
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.