Learn More
Selleck Chemical LLC Sunitinib 200mg 557795-19-4 SU11248
SDP
Supplier:  Selleck Chemical LLC S7781200MG
Encompass_Preferred
Sunitinib (SU11248) is a multi-targeted RTK inhibitor targeting VEGFR2 (Flk-1) and PDGFR? with IC50 of 80 nM and 2 nM, and also inhibits c-Kit. Sunitinib is also a dose-dependent inhibitor of the autophosphorylation activity of IRE1?. Sunitinib induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5240
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.