Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sunitinib malate

Catalog No. AC462640010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462640010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC462640010 Supplier Thermo Scientific Chemicals Supplier No. 462640010

missing translation for 'validMainframeMsg'

CAS: 341031-54-7 | C26H33FN4O7 | 532.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 341031-54-7
Molecular Formula C26H33FN4O7
Molecular Weight (g/mol) 532.57
InChI Key LBWFXVZLPYTWQI-IPOVEDGCSA-N
IUPAC Name (2S)-2-hydroxybutanedioic acid; N-[2-(diethylamino)ethyl]-5-{[(3Z)-5-fluoro-2-oxo-2,3-dihydro-1H-indol-3-ylidene]methyl}-2,4-dimethyl-1H-pyrrole-3-carboxamide
SMILES O[C@@H](CC(O)=O)C(O)=O.CCN(CC)CCNC(=O)C1=C(C)NC(\C=C2/C(=O)NC3=CC=C(F)C=C23)=C1C

Specifications

Color Clear yellow to yellow-orange solution
Quantity 1 g
Formula Weight 532.56
Chemical Name or Material Sunitinib malate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement May damage fertility or the unborn child.

Causes damage to organs through prolonged or repeated exposure if inhaled.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.