accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH3CHNOH3CNHOHNFCH3OH(S)HOOOHO(Z)

CAS RN 341031-54-7

IUPAC Name: (2S)-2-hydroxybutanedioic acid; N-[2-(diethylamino)ethyl]-5-{[(3Z)-5-fluoro-2-oxo-2,3-dihydro-1H-indol-3-ylidene]methyl}-2,4-dimethyl-1H-pyrrole-3-carboxamide
Synonyms: sunitinib
Molecular Weight (g/mol): 532.57
Molecular Formula: C26H33FN4O7
InChi Key: LBWFXVZLPYTWQI-IPOVEDGCSA-N
SMILES: O[C@@H](CC(O)=O)C(O)=O.CCN(CC)CCNC(=O)C1=C(C)NC(\C=C2/C(=O)NC3=CC=C(F)C=C23)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)