Description
Supel™Flash Cartridges are pre-compressed, disposable flash columns containing ultrapure silica, designed to meet your stringent flash purification requirements. Cartridge pre-compression reduces particle interstitial spacing to provide even mass transfer and balanced chemical interaction.
Features Include:
- Female luer lock inlet and male slip luer outlet for easy compatibility
- 8 Bed weights from 4 grams to 330 grams
- 40-60 μm irregular silica
- Higher efficiency leading to sharper more symmetric peaks
- High yield and sample loading capacity
- Reliable, consistent purification performance
- Supel Flash Cartridges are Designed For Use in: ISCO's CombiFlash, Companion, Retrieve, and OptiX systems; Analogix's SimpliFlash, and IntelliFlash and Biotage systems.
Specifications
Specifications
| Includes | 15/Pk. |
| Bed Weight (Metric) | 40 g |
| Column Type | Flash Chromatography Cartridge |
| For Use With (Application) | Flash Chromatography |
| For Use With (Equipment) | Isco and Analogix Flash Systems |
| Product Line | Supel |
Supel is a trademark of Sigma-Aldrich Co. LLC