Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Supel™ Swift HLB SPE 96-Well Plate

The 96-well format is perfect for medium to high-throughput laboratories performing large numbers of samples at a time and is also amenable to automation.

$388.00

Specifications

Particle Size 40 to 75 μm
Matrix Polymeric HLB Base Material
Pore Size 80 to 210 Å
Quantity 1 Ea.
Surface Area (Metric) 425 to 500 m2/g
View More Specs

Products 2
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
11-102-3076
View Documents
MilliporeSigma™ Supelco™
57495U
10 mg
Each for $388.00
Only null left
Add to Cart
 
11-102-3075
View Documents
MilliporeSigma™ Supelco™
57494U
30 mg
Each for $388.00
Only null left
Add to Cart
 
Description

Description

  • Supel™ Swift HLB is a co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups.
  • This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma).
  • Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values.
  • Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions.
  • The product design allows for direct sample loading onto the sorbent bed, thereby eliminating the conditioning step altogether.
  • This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.
  • The 96-well format is perfect for medium to high-throughput laboratories performing large numbers of samples at a time and is also amenable to automation.
Specifications

Specifications

40 to 75 μm
80 to 210 Å
425 to 500 m2/g
Polymeric HLB Base Material
1 Ea.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.