Learn More
Supel™ Swift HLB SPE Tubes, Sold by MilliporeSigma™ Supelco™
Catalog No. 111022232 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Model:
200 mg (bed)
30 mg (bed)
60 mg (bed)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
11-102-2232 30 mg (bed)
11-102-2230 200 mg (bed)
11-102-2231 60 mg (bed)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-102-2232 Supplier MilliporeSigma Supelco Supplier No. 57493U
Add to Cart
Add to Cart
Co-polymer solid phase extraction (SPE) phase composed of both hydrophilic and lipophilic functional groups.
  • This phase is ideal for extracting a broad range of compounds from aqueous samples, including food and environmental samples, as well as biological samples (eg. serum and plasma).
  • Because of the hydrophilic and lipophilic balanced (HLB) property, the polymer material can selectively retain a wide range of compounds with different polarities and LogP values.
  • Traditional HLB SPE uses a five-step process for sample preparation that normally includes the conditioning of sorbents with an organic solvent followed by an equilibration step to return sorbents to sample loading conditions. The product design allows for direct sample loading onto the sorbent bed, thereby eliminating conditioning of the cartridge. This product feature can both save time and simplify sample preparation, which may also serve to reduce sample processing errors.
TRUSTED_SUSTAINABILITY
Specifications
Type 80 to 210 Å Pore Size
Includes 40-75 μm (>90%) particle size
Model 30 mg (bed)
Volume (Metric) 1 mL
Description Detection MS friendly
Supel is a trademark of Sigma-Aldrich Co. LLC