Learn More
MilliporeSigma™ Supelco™ Supelco Ferrule Nut Adapter (fits Agilent injectors and non-MS detectors), hexagonal wrenchtight version
Catalog No. 111011553 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Packaging Quantity:
2/Pk.
Catalog No. Packaging Quantity
11-101-1553 2/Pk.
1 options
Catalog No. 11-101-1553 Supplier MilliporeSigma™ Supelco™ Supplier No. 22470U
Add to Cart
Add to Cart
These stainless steel adapters enable you to use general purpose and other 1/16 in. compression ferrules in Agilent GCs. Hexagonal shape allows tightening with a wrench to keep fingers from getting burned.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Quantity 2/Pk.
For Use With Agilent Injectors and Non-MS Detectors